Learn More
BEST SANITIZERS Alpet D2 Quat-Free Surface Sanitizer, Quart
Supplier:  BEST SANITIZERS SS10032
Encompass_Preferred
This item has a minimum qty of 2 per supplier requirements.
Alpet D2 Quat-Free Surface Sanitizer is a ready-to-use, no-rinse, fast acting, food contact and non-food contact surface cleaner and sanitizer. It is highly evaporative and ideal for water sensitive equipment and dry environments. It leaves no unsightly residue, and is an effective sanitizer for those processors who can't use quat.Clean, Sanitize and Disinfect food contact and non-food contact surfaces with one productReady-to-Use, no-rinse required70% Alcohol Formula (62.5% ethanol / 7.5% isopropanol)Alpet D2 Quat-Free Surface Sanitizer is NSF listed, Kosher, Pareve and Halal certified60 Second Kill on food contact surfaces, including rubber gloves*10 Second Kill on non-food contact surfaces, including rubber footwear
Catalog No. NC1479486