Learn More
CSAFE LLC CSafe Global 6L Refrigerated Courier Solution for 2°C to 8°C. Maintains 2°C to 8°C for 12 hours.
Catalog No. NC1726161
Encompass
CSafe Courier solutions are designed for physicians, pharmacists, researchers, pharmaceutical representatives, and other healthcare professionals making last-mile delivery of temperature-sensitive life science products. Maintains temperature for 12 hours.Product Storage Temperature: 2°C to 8°C (36°F to 46°F) | Payload Volume: 8.2 L (512in³) | Internal Payload Dimensions mm (inches): 203 x 203 x 203 (8 x 8 x 8).
This item is not returnable. View return policy
Catalog No. NC1726161 Supplier CSAFE LLC Supplier No. PX6LS28
CSafe Courier solutions are designed for physicians, pharmacists, researchers, pharmaceutical representatives, and other healthcare professionals making last-mile delivery of temperature-sensitive life science products. Maintains temperature for 12 hours.Product Storage Temperature: 2°C to 8°C (36°F to 46°F) | Payload Volume: 8.2 L (512in³) | Internal Payload Dimensions mm (inches): 203 x 203 x 203 (8 x 8 x 8).