Learn More
Cole-Parmer Fowler 54-100-112-1 12" Digital Caliper with LCD - +/-0.0015 in/+/-0.04 mm
Supplier:  Cole-Parmer 9715212
Encompass_Preferred
This digital caliper features a unique ceramic capacitive sensor. Measure inside, outside, depth, step, with direct inch/metric conversion and 0.0005"/0.01 mm resolution.
Catalog No. NC9534930