Learn More
Ace Glass, Inc. 12L complete reactor assembly. Tall support raises mantle36" above the floor.
SDP
Catalog No. 50978751
Encompass
12L complete reactor assembly includes flask, self-standing mechanical or air motor, flat-sided powder funnel, slow speed operation motor control, rugged steel reinforced PTFE stirring rod, and a tall mantle support. This complete self-supporting pilot plant assembly needs only to be located near coolant and electrical outlet. When not in use the entire assembly can be intact without breakdown.
Catalog No. 50-978-751 Supplier Ace Glass, Inc. Supplier No. 646962
Add to Cart
Add to Cart
Request a Quote
12L complete reactor assembly includes flask, self-standing mechanical or air motor, flat-sided powder funnel, slow speed operation motor control, rugged steel reinforced PTFE stirring rod, and a tall mantle support. This complete self-supporting pilot plant assembly needs only to be located near coolant and electrical outlet. When not in use the entire assembly can be intact without breakdown.