Learn More
Chemglass Life Sciences 12L Globe Shaped Separatory Funnel, Large Capacity, #45/50 Outer Stopper Neck, 10mm PTFE Stpk
SDP
Catalog No. 5000980634
Encompass_Preferred
Large capacity globe shaped separatory funnel with a choice of glass or PTFE stopcock. Stem length is approx. 130mm below the stopcock on all sizes. For replacement stoppers see CG-3018 for #38, CG-3000 for 45/50.
Catalog No. 50-009-80634 Supplier Chemglass Life Sciences Supplier No. CG173906
Large capacity globe shaped separatory funnel with a choice of glass or PTFE stopcock. Stem length is approx. 130mm below the stopcock on all sizes. For replacement stoppers see CG-3018 for #38, CG-3000 for 45/50.