Learn More
Chemglass Life Sciences 12000mL 3-Neck Round Bottom Flask (RBF) , Half Jacket, 1-CN 45/50 Outer, 2-SN 24/40 Outer, 12mm Drain Valve
SDP
Catalog No. CG153911
Encompass_Preferred
12000mL 3-Neck RBF, Half Jacket, 1-CN 45/50 Outer, 2-SN 24/40 Outer, 12mm Drain Valve. Lower half of flask has a jacket permitting heating or cooling of the contents with fluid from a constant temperature bath.The design of the Chem-Drain valve incorporates a PTFE plunger that allows the contents of the flask to drain directly through the bore of the plunger. When closed, the tip of the PTFE plug seals into the lower radius of the flask, avoiding the dead space normally associated with standard stopcock drains. This feature permits total mixing and drainage of the flask's contents. The Chem-Drain is 12mm. This dimension refers to the straight section of the plunger.
Catalog No. CG153911 Supplier Chemglass Life Sciences Supplier No. CG153911
12000mL 3-Neck RBF, Half Jacket, 1-CN 45/50 Outer, 2-SN 24/40 Outer, 12mm Drain Valve. Lower half of flask has a jacket permitting heating or cooling of the contents with fluid from a constant temperature bath.The design of the Chem-Drain valve incorporates a PTFE plunger that allows the contents of the flask to drain directly through the bore of the plunger. When closed, the tip of the PTFE plug seals into the lower radius of the flask, avoiding the dead space normally associated with standard stopcock drains. This feature permits total mixing and drainage of the flask's contents. The Chem-Drain is 12mm. This dimension refers to the straight section of the plunger.