Learn More
Chemglass Life Sciences 12000ml Reaction Vessel, Spherical, 150mm Schott O-Ring Flange, 295mm O.D. x 365mm Height
SDP
Catalog No. CG193204
Encompass_Preferred
12000ml Reaction Vessel, Spherical, 150mm Schott O-Ring Flange, 295mm O.D. x 365mm Height. Spherical shaped reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Vessel will fit standard heating mantles CG-10000 through CG-12502. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.
Catalog No. CG193204 Supplier Chemglass Life Sciences Supplier No. CG193204
12000ml Reaction Vessel, Spherical, 150mm Schott O-Ring Flange, 295mm O.D. x 365mm Height. Spherical shaped reaction vessel constructed with a flange having a groove formed in the sealing surface of the flange. Flange is also ground to permit a greased seal with any of the lids CG-1940 through CG-1946. Vessel will fit standard heating mantles CG-10000 through CG-12502. For clamp see CG-141 or CG-141-T. For o-ring see CG-147.