Learn More
AirSea USA 12L UN Steel Drum, 1A2/Y1.8/200 & 1A2/X28/S
Catalog No. 501534813
Encompass_Preferred
12L UN Steel drum that can be placed in code 424 to ship large amounts of UN2814 infectious substances. UN Marks: 1A2/Y1.8/200 & 1A2/X28/S. 12L UN Steel Drum With Lacquer Lining & Lever Locking Ring. Approved to ship both solid and liquid dangerous goods substances.
Catalog No. 50-153-4813 Supplier AirSea USA Supplier No. 437
12L UN Steel drum that can be placed in code 424 to ship large amounts of UN2814 infectious substances. UN Marks: 1A2/Y1.8/200 & 1A2/X28/S. 12L UN Steel Drum With Lacquer Lining & Lever Locking Ring. Approved to ship both solid and liquid dangerous goods substances.