Learn More
Trajan Scientific 12M X 0.22MM ID - BP1 0.25UM
SDP
Catalog No. 502530245
Encompass_Preferred
Trajan BP1 GC Capillary Columns, Diameter Inner: 0.22 mm, Film Thickness: 0.25 mm, Length: 12 mm, Stationary Phase: 100% Dimethyl Polysiloxane, Max. Temperature: 360 deg.C, Quantity: 1 Each, Temperature Range: -60 deg.C to +360 deg.C
Catalog No. 50-253-0245 Supplier Trajan Scientific Supplier No. 054046
Trajan BP1 GC Capillary Columns, Diameter Inner: 0.22 mm, Film Thickness: 0.25 mm, Length: 12 mm, Stationary Phase: 100% Dimethyl Polysiloxane, Max. Temperature: 360 deg.C, Quantity: 1 Each, Temperature Range: -60 deg.C to +360 deg.C