Learn More
Cayman Chemical 12methyl TrIDcnoIc ACD me 10mg
SDP
Catalog No. 503015835
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in beef tallow, mutton tallow, and cod liver oil; has been used as a standard for the quantification of 12-methyl tridecanoic acid in the triglyceride fraction of skin surface lipids from human males by GC-MS
Catalog No. 50-301-5835 Supplier Cayman Chemical Supplier No. 2481010 MG
A methylated fatty acid methyl ester; has been found in beef tallow, mutton tallow, and cod liver oil; has been used as a standard for the quantification of 12-methyl tridecanoic acid in the triglyceride fraction of skin surface lipids from human males by GC-MS