Learn More
Cayman Chemical 12-methyl Tridecanoic Acid methyl ester, Purity: ≥98%, CAS: 5129-58-8, 20 mg
SDP
Catalog No. 503016004
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in beef tallow, mutton tallow, and cod liver oil; has been used as a standard for the quantification of 12-methyl tridecanoic acid in the triglyceride fraction of skin surface lipids from human males by GC-MS
Catalog No. 50-301-6004 Supplier Cayman Chemical Supplier No. 2481020 MG
A methylated fatty acid methyl ester; has been found in beef tallow, mutton tallow, and cod liver oil; has been used as a standard for the quantification of 12-methyl tridecanoic acid in the triglyceride fraction of skin surface lipids from human males by GC-MS