Learn More
Chemglass Life Sciences 12mm Control Knob Only
SDP
Catalog No. 501531787
Encompass_Preferred
12mm Control Knob Only. Replacement 12mm Chem-Cap stem and control knob for Chemglass cylindrical and spherical vessels. PTFE stem is sold complete with Perfluoro or Viton o-rings installed under each Chem-Cap and one installed as the back up o-ring.
Catalog No. 50-153-1787 Supplier Chemglass Life Sciences Supplier No. CG51130
12mm Control Knob Only. Replacement 12mm Chem-Cap stem and control knob for Chemglass cylindrical and spherical vessels. PTFE stem is sold complete with Perfluoro or Viton o-rings installed under each Chem-Cap and one installed as the back up o-ring.