Learn More
Chemglass Life Sciences 12mm Extended Tip Drain Valve, Complete
SDP
Catalog No. CG51101
Encompass_Preferred
12mm Extended Tip Drain Valve, Complete. Designed to be used as a drain for reaction kettles, flasks, etc. Valve is made with an extended tip which fills the seat area and permits total agitation of the contents of the vessel. Once opened, contents of the vessel will drain through the sidearm. Valve is constructed the same as the rest of our Chem-Cap valve line, and has an integral PTFE tip o-ring. Supplied complete PTFE stem with Viton o-rings, control knob, and corresponding glass barrel having a single sidearm which can be further fabricated into the drain arm. PLEASE NOTE: COMPLETE ITEM IS SUPPLIED WITH A 90 1-ARM GLASS VALVE BODY.
Catalog No. CG51101 Supplier Chemglass Life Sciences Supplier No. CG51101
12mm Extended Tip Drain Valve, Complete. Designed to be used as a drain for reaction kettles, flasks, etc. Valve is made with an extended tip which fills the seat area and permits total agitation of the contents of the vessel. Once opened, contents of the vessel will drain through the sidearm. Valve is constructed the same as the rest of our Chem-Cap valve line, and has an integral PTFE tip o-ring. Supplied complete PTFE stem with Viton o-rings, control knob, and corresponding glass barrel having a single sidearm which can be further fabricated into the drain arm. PLEASE NOTE: COMPLETE ITEM IS SUPPLIED WITH A 90 1-ARM GLASS VALVE BODY.