Learn More
Chemglass Life Sciences Fritted Disc, 12mm, Extra Coarse
SDP
Catalog No. 501531957
Encompass_Preferred
Fritted Disc, 12mm, Extra Coarse. Porous discs for use in fabrication of special glass apparatus. Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI. Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.
Catalog No. 50-153-1957 Supplier Chemglass Life Sciences Supplier No. CGB20108
Fritted Disc, 12mm, Extra Coarse. Porous discs for use in fabrication of special glass apparatus. Fritted glassware is designed for low pressure vacuum filtration or gas flow. The MAX differential pressure should not exceed 15 PSI. Fritted glassware should not be exposed to excessive temperature change or direct exposure to flame due to the fact that it has less resistance to thermal shock than standard borosilicate glassware.