Learn More
Ace Glass, Inc. 12MM GRADED SEALS 12MM GRADED SEALS
SDP
Supplier:  Ace Glass, Inc. 845520
Encompass
12mm quartz graded seals for connecting Pyrex or Kimax borosilicate glass to Quartz glass. Quartz length is 7.6cm, and borosilicate length is 10.2cm.
Catalog No. 50-974-596