Learn More
Glas Col LLC Evaporator Accessory
SDP
Supplier:  Glas Col LLC 109A YH2412
Encompass_Preferred
Aluminum insert block, 24 position for up to 12mm vials. Use with small evpaorator system 109A YH-1.
Catalog No. NC1285038