Learn More
MTC Bio 26' Cassette of 12mm lab tape, white w/ black print
Catalog No. NC1334014
Encompass_Preferred
This item has a minimum qty of 6 per supplier requirements.
The new LABeler™ Lab Printer provides the convenience of a lab pen with the benefits of a large desktop printer. Simply type in the desired copy using the 50+ button key pad and press the print key within seconds your label can be cut by pressing the cut lever and is then ready for application. With over 15 available sizes, colors & material combinations, the LABeler is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50C. Special Ultra-Low label tapes are also available for storage in -80C environments or for liquid nitrogen applications.
Catalog No. NC1334014 Supplier MTC Bio Supplier No. L901012WK
This item has a minimum qty of 6 per supplier requirements.
The new LABeler™ Lab Printer provides the convenience of a lab pen with the benefits of a large desktop printer. Simply type in the desired copy using the 50+ button key pad and press the print key within seconds your label can be cut by pressing the cut lever and is then ready for application. With over 15 available sizes, colors & material combinations, the LABeler is ideal for label identification of any laboratory vessel for nearly any laboratory application. Standard label tapes can be autoclaved, boiled or even frozen to temperatures as low as -50C. Special Ultra-Low label tapes are also available for storage in -80C environments or for liquid nitrogen applications.