Learn More
Chemglass Life Sciences 12mm Probe Holder, Fixed Height, G3/4 Thread
SDP
Catalog No. 501294437
Encompass
12mm Probe Holder, Fixed Height, G3/4 Thread. Probe holders for 12mm probes. These are non-height adjustable.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
This item is not returnable. View return policy
Catalog No. 50-129-4437 Supplier Chemglass Life Sciences Supplier No. CLS138109
12mm Probe Holder, Fixed Height, G3/4 Thread. Probe holders for 12mm probes. These are non-height adjustable.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.