Learn More
Chemglass Life Sciences 12mm Probe Holder, Fixed Height, M18 Thread
SDP
Catalog No. 501294493
Encompass
12mm Probe Holder, Fixed Height, M18 Thread. Probe holders for 12mm probes. These are non-height adjustable.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.
This item is not returnable. View return policy
Catalog No. 50-129-4493 Supplier Chemglass Life Sciences Supplier No. CLS138108
12mm Probe Holder, Fixed Height, M18 Thread. Probe holders for 12mm probes. These are non-height adjustable.Like-for-like replacement parts for Applikon Biotechnology and Broadley-James brands of bioreactors.Applikon is a registered trademark of Applikon Biotechnology, Inc. Broadley-James and Broadley-James Corporation are registered trademarks of Broadley-James Corporation.