Learn More
Chemglass Life Sciences Chemglass 12 mm PTFE Stem, Replacement for Chem-Drain Valve
SDP
Catalog No. 502733522
Encompass_Preferred
Replacement PTFE stem for 12 mm drain valves, intended for use as a spare or repair part to restore sealing and flow control in Chem-Drain style valve assemblies. Manufactured from PTFE for broad chemical resistance and smooth operation, the stem is sized to fit 12 mm valve assemblies and permits drainage of particulate-laden solutions.
  • 12 mm stem diameter for standard 12 mm drain valve assemblies.
  • PTFE material provides excellent chemical resistance and low friction.
  • Permits drainage of liquids and solids up to 2 mm in diameter.
  • Designed for use as a replacement or spare stem for repair and maintenance.
  • Simple installation; compatible with valve bodies where PTFE components are supported.
Catalog No. 50-273-3522 Supplier Chemglass Life Sciences Supplier No. CG51020
Replacement PTFE stem for 12 mm drain valves, intended for use as a spare or repair part to restore sealing and flow control in Chem-Drain style valve assemblies. Manufactured from PTFE for broad chemical resistance and smooth operation, the stem is sized to fit 12 mm valve assemblies and permits drainage of particulate-laden solutions.
  • 12 mm stem diameter for standard 12 mm drain valve assemblies.
  • PTFE material provides excellent chemical resistance and low friction.
  • Permits drainage of liquids and solids up to 2 mm in diameter.
  • Designed for use as a replacement or spare stem for repair and maintenance.
  • Simple installation; compatible with valve bodies where PTFE components are supported.