Learn More
Neuvitro Laminin coated sterile German glass coverslips for cell culture, 12mm dia #1 thick 60/pk, ready to use in cell culture.
Supplier:  Neuvitro GG12LAMINN
Encompass_Preferred
CE certified German glass coverslips with Laminin coating on both sides for cell culture and confocal / IHC imaging with no autofluorescence. Pre-washed, nitric acid treated, coated with pathgen-free mouse laminin on both sides and sterile. 12mm diameter round, #1 thickness, 60 pieces/pack. Ready to use directly in cell culture. Coverslips are sealed as 15 pieces per group for easy storage and enhanced cell culture quality. Shelf life 0.5 year when stored in chest freezer. User manual included.
Catalog No. NC0634548