Learn More
Electron Microscopy Sciences AFM Metal Specimen Discs, 12 mm
SDP
Catalog No. 5033080
Encompass_Preferred
These are high quality metal discs with smooth edges and flat surfaces for use in Atomic force Microscopy.
Diameter: 12 mm
Catalog No. 50-330-80 Supplier Electron Microscopy Sciences Supplier No. 7501012
These are high quality metal discs with smooth edges and flat surfaces for use in Atomic force Microscopy.
Diameter: 12 mm