Learn More
Waters Corp Waters Vial Collection Rack, 12 mm, Holds 24 Vials, Consumable
Supplier:  Waters Corp 186010530
Encompass_Preferred
The 12 mm Vial Collection Rack is a consumable accessory designed to organize and hold 24 vials. It ensures efficient sample handling and is engineered for seamless integration within laboratory automation workflows.
  • Designed to hold 24 vials.
  • Compatible with the Extraction+ Device.
  • Compatible with the Collection Rack Domino.
  • Can be moved by the Andrew+ robot using the Microplate Gripper tool.
  • Consumable rack for easy replacement.
Catalog No. 50-226-9416