Learn More
Ace Glass, Inc. PTFE 12mm tube compression fitting to 1/2in male NPT
SDP
Catalog No. 50977686
Encompass
12mm PTFE tube compression fitting to 1/2 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-686 Supplier Ace Glass, Inc. Supplier No. 1270950
12mm PTFE tube compression fitting to 1/2 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.