Learn More
Ace Glass, Inc. PTFE 12mm tube compression fitting to 3/8in male NPT
SDP
Catalog No. 50977683
Encompass
12mm PTFE tube compression fitting to 3/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.
Catalog No. 50-977-683 Supplier Ace Glass, Inc. Supplier No. 1270942
12mm PTFE tube compression fitting to 3/8 male NPT. All wetted surfaces are virgin PTFE. Nuts and gripper are PVDF.