Learn More
Cayman Chemical 12-methyl Myristic Acid methyl ester, Purity: ≥98%, CAS: 5129-66-8, 5 mg
SDP
Catalog No. 503015860
Encompass_Preferred
A methylated fatty acid methyl ester; has been found in vermicomposts of cattle manure, carica papaya leaves, and cuticular wax of K. africana; a volatile compound in lipid-lowering granulated tea; levels are decreased in nifurtimox-treated T. cruzi compared to non-treated controls
Catalog No. 50-301-5860 Supplier Cayman Chemical Supplier No. 248185 MG
A methylated fatty acid methyl ester; has been found in vermicomposts of cattle manure, carica papaya leaves, and cuticular wax of K. africana; a volatile compound in lipid-lowering granulated tea; levels are decreased in nifurtimox-treated T. cruzi compared to non-treated controls