Learn More
Cayman Chemical 12-oxo Stearic Acid methyl ester, Purity: ≥95%, CAS: 2380-27-0, 250 mg
SDP
Catalog No. 502888943
Encompass_Preferred
An esterified form of 12-oxo stearic acid; has been found in esterified L. chinensis and L. decipiens pulp oils and serves as a measure of 12-oxo stearic acid content
Catalog No. 50-288-8943 Supplier Cayman Chemical Supplier No. 10007012250 MG
An esterified form of 12-oxo stearic acid; has been found in esterified L. chinensis and L. decipiens pulp oils and serves as a measure of 12-oxo stearic acid content