Learn More
Cayman Chemical 12-oxo Stearic Acid methyl ester, Purity: ≥95%, CAS: 2380-27-0, 50 mg
SDP
Catalog No. 502887894
Encompass_Preferred
An esterified form of 12-oxo stearic acid; has been found in esterified L. chinensis and L. decipiens pulp oils and serves as a measure of 12-oxo stearic acid content
Catalog No. 50-288-7894 Supplier Cayman Chemical Supplier No. 1000701250 MG
An esterified form of 12-oxo stearic acid; has been found in esterified L. chinensis and L. decipiens pulp oils and serves as a measure of 12-oxo stearic acid content