Learn More
Nordson EFD 12 fl oz (360 ml) Clear Optimum Cartridges, 25 per box
SDP
Catalog No. NC1626692
Encompass
Molded from proprietary FDA- and RoHS-compliant polymer, Optimum cartridges provide exceptional clarity, chemical compatibility, impact strength, and dimensional stability. The Zero Draft™ design improves fluid flow characteristics and reduces turbulence by ensuring the internal diameter is consistent from top to bottom with no taper restricting piston travel or affecting process results. Unique external ribs at the outlet end prevent splitting when installing fittings or nozzles. Precision molded threads and a tapered counterbore at the 0.25” NPT outlet create a secure, leakproof seal.
Catalog No. NC1626692 Supplier Nordson EFD Supplier No. 7012407
Molded from proprietary FDA- and RoHS-compliant polymer, Optimum cartridges provide exceptional clarity, chemical compatibility, impact strength, and dimensional stability. The Zero Draft™ design improves fluid flow characteristics and reduces turbulence by ensuring the internal diameter is consistent from top to bottom with no taper restricting piston travel or affecting process results. Unique external ribs at the outlet end prevent splitting when installing fittings or nozzles. Precision molded threads and a tapered counterbore at the 0.25” NPT outlet create a secure, leakproof seal.