Learn More
TERRISS INDUSTRIES INC Certified Control Drink Flask 12 oz
Catalog No. NC0351233
Encompass
These Control Drink Flasks are designed to equate, in terms of capacity, an empty, dry container. The flasks, when properly filled, will hold the exact same content as a bottle and can be used to check proportioning.Calibrated in 1/10 or 1 ml Divisions.
Catalog No. NC0351233 Supplier TERRISS INDUSTRIES INC Supplier No. T09110
These Control Drink Flasks are designed to equate, in terms of capacity, an empty, dry container. The flasks, when properly filled, will hold the exact same content as a bottle and can be used to check proportioning.Calibrated in 1/10 or 1 ml Divisions.