Learn More
C&G Containers 12OZ PRE CLEANED MAYO BTL 12CS
SDP
Catalog No. NC9710045
Encompass_Preferred
12oz Flint glass Mayo 70-450G White PP cap PTFE-lined, certified B1/Barcoded/Labels 12/cs
Catalog No. NC9710045 Supplier C&G Containers Supplier No. LFM0124000
12oz Flint glass Mayo 70-450G White PP cap PTFE-lined, certified B1/Barcoded/Labels 12/cs