Learn More
USA Scientific Inc 12-place chest freezer rk
Catalog No. 502744882
Encompass
12-place (1 long x 12 high) chest freezer rack for 2 in H boxes, 5 1/2 in L x 5 5/8 in W x 26 3/8 in H, stainless steel
Catalog No. 50-274-4882 Supplier USA Scientific Inc Supplier No. 26021200
12-place (1 long x 12 high) chest freezer rack for 2 in H boxes, 5 1/2 in L x 5 5/8 in W x 26 3/8 in H, stainless steel