Learn More
USA Scientific Inc 12-place upright freezer rk
Catalog No. 502745083
Encompass
12-place (4 long x 3 high) upright freezer rack for 3 in H boxes, 22 1/8 in L x 5 1/2 in W x 9 5/8 in H, stainless steel
Catalog No. 50-274-5083 Supplier USA Scientific Inc Supplier No. 26034300
12-place (4 long x 3 high) upright freezer rack for 3 in H boxes, 22 1/8 in L x 5 1/2 in W x 9 5/8 in H, stainless steel