Learn More
USA Scientific Inc 12-place upright frzr rk
Catalog No. 502745003
Encompass
12-place (3 long x 4 high)eco-design upright freezer rack for 2 in H boxes, 16 5/16 in L x 5 1/2 in W x 8 7/8 in H, stainless steel
Catalog No. 50-274-5003 Supplier USA Scientific Inc Supplier No. 26233400
12-place (3 long x 4 high)eco-design upright freezer rack for 2 in H boxes, 16 5/16 in L x 5 1/2 in W x 8 7/8 in H, stainless steel