Learn More
Alkali Scientific 12-Strip Tubes and 12-Strip Flat Caps, Packed Together, 80 Strips/pk
SDP
Catalog No. 502336188
Encompass_Preferred
Angularly attached caps prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack.Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing. Clear tubes provide high clarity for visualizing contents. For use in pharmaceutical industry, food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.
Catalog No. 50-233-6188 Supplier Alkali Scientific Supplier No. PC7014
Angularly attached caps prevent cap lids and hinges from interfering with each other, allowing for easier operation in an SBS format rack.Ultra-thin walls ensure efficient thermal transfer and maximum yield, yet are strong enough to prevent crushing. Clear tubes provide high clarity for visualizing contents. For use in pharmaceutical industry, food industry, and molecular biology. Manufactured from high-quality, virgin polypropylene to ensure purity and provide excellent optical clarity. Certified free of detectable RNase, DNase, DNA, PCR inhibitors, and tested pyrogen-free.