Learn More
Hach Company 12V Lead Acid Batt 3 pin conn.
Supplier:  Hach Company 8754400
Encompass
12 V lead acid battery with 3-pin connector for SD900/AS950 sampler Use with charger 8753500
Catalog No. 50-002-07426