Learn More
USA Scientific Inc 12-w round bottom reservoir
Catalog No. 502745433
Encompass
Twelve well polypropylene reservoir with lid, individually wrapped, str, 20/pack. 5 mL/well.
Catalog No. 50-274-5433 Supplier USA Scientific Inc Supplier No. 13011212
Twelve well polypropylene reservoir with lid, individually wrapped, str, 20/pack. 5 mL/well.