Learn More
Chemglass Life Sciences 12 Well Cell Culture Plate, Flat, Non-Treated, Sterile, 1/pk, 50/cs
SDP
Catalog No. 501897684
Encompass_Preferred
Nest cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform growth. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Nontreated growth surface is more hydrophobic than tissue culture surface and is similar to the surface of a bacteriological Petri dish.Made from high clarity, 100% virgin polystyreneSterilized by E-beamClear lot number for batch traceabilityPremium packaging; Individually Wrapped in Peel-able Tyvek Films and BlistersNon-Pyrogenic, DNase/RNase-Free
Catalog No. 50-189-7684 Supplier Chemglass Life Sciences Supplier No. CGN3501212
Add to Cart
Add to Cart
Nest cell culture plates have round edges for better handling. Low profile, low-evaporation lid design assures uniform growth. Small feet on the bottom corners of each plate eliminate scraping of the microscope lens. Nontreated growth surface is more hydrophobic than tissue culture surface and is similar to the surface of a bacteriological Petri dish.Made from high clarity, 100% virgin polystyreneSterilized by E-beamClear lot number for batch traceabilityPremium packaging; Individually Wrapped in Peel-able Tyvek Films and BlistersNon-Pyrogenic, DNase/RNase-Free