Learn More
Altenloh Brinck & Co US INC. DBA: SABEU 12 well insert 0.4um PET 4Plate (cellQART)
Catalog No. 502673507
Encompass_Preferred
cellQART Cell Culture Inserts combined with our especially designed Well Plates are intended to be used for cell culture and in vitro tissue culture research applications. The Well Plate is supplied in 12-Well format, it is sterile and fully pre-loaded with cellQART Cell Culture Inserts. These cellQART Cell Culture Inserts come with PET membranes in clear optics. The Inserts are tissue culture treated and sterile. They are ideal for in-situ phase contrast microscopy. Longer studies/testing (2weeks+) and imaging clarity is best suited
Catalog No. 50-267-3507 Supplier Altenloh Brinck & Co US INC. Supplier No. 9310424
cellQART Cell Culture Inserts combined with our especially designed Well Plates are intended to be used for cell culture and in vitro tissue culture research applications. The Well Plate is supplied in 12-Well format, it is sterile and fully pre-loaded with cellQART Cell Culture Inserts. These cellQART Cell Culture Inserts come with PET membranes in clear optics. The Inserts are tissue culture treated and sterile. They are ideal for in-situ phase contrast microscopy. Longer studies/testing (2weeks+) and imaging clarity is best suited