Learn More
USA Scientific Inc 12-wl v-bottom reservoir
Catalog No. 502745016
Encompass
Twelve well v-bottom reservoir with lid, individually wrapped, 50/pack. Each v-shaped well holds up to 3 mL.
Catalog No. 50-274-5016 Supplier USA Scientific Inc Supplier No. 38233412
Twelve well v-bottom reservoir with lid, individually wrapped, 50/pack. Each v-shaped well holds up to 3 mL.