Learn More
Chemglass Life Sciences Plate glass, square, borosilicate, 12in X 12in X 1/16
SDP
Catalog No. CG190442
Encompass_Preferred
Plate glass, square, borosilicate, 12in X 12in X 1/16. Pyrex or Duran borosilicate plate glass. The 1/16in, 1/8in and 1/4in thick plates have edges that are cut and swiped. The 1/2in thick and over have ground edges.
Catalog No. CG190442 Supplier Chemglass Life Sciences Supplier No. CG190442
Plate glass, square, borosilicate, 12in X 12in X 1/16. Pyrex or Duran borosilicate plate glass. The 1/16in, 1/8in and 1/4in thick plates have edges that are cut and swiped. The 1/2in thick and over have ground edges.