Learn More
FUMEX INC FA1 Mini Gas/Odor Filter
SDP
Catalog No. NC1729456
Encompass
The 12” x 12” x 2” FA101D 3D printer filter & laser printer filter incorporates high performance bonded activated carbon technology designed for the removal of the gases/odors produced in 3D printing & laser applications. Bonded technology provides superior adsorption with minimal airflow resistance.
Catalog No. NC1729456 Supplier FUMEX INC Supplier No. FA101D
The 12” x 12” x 2” FA101D 3D printer filter & laser printer filter incorporates high performance bonded activated carbon technology designed for the removal of the gases/odors produced in 3D printing & laser applications. Bonded technology provides superior adsorption with minimal airflow resistance.