Learn More
Chemglass Life Sciences Graded Seal, Quartz to Borosilicate, 12mm ID X 14mm OD
SDP
Catalog No. CGQ031011
Encompass_Preferred
Graded Seal, Quartz to Borosilicate, 12mm ID X 14mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.
Catalog No. CGQ031011 Supplier Chemglass Life Sciences Supplier No. CGQ031011
Graded Seal, Quartz to Borosilicate, 12mm ID X 14mm OD. Graded seals have 3 inches of quartz sealed to 4 inches of borosilicate.