Learn More
MSC Koch Filter OdorKleen ES, 12x24x4 in, MERV-8, 6/Case
Catalog No. 5000506779
Encompass
This extended surface carbon / particulate pleated panel filter is designed for superior filtration in commercial HVAC systems. It utilizes specialized, 100% high-efficiency mechanical media to effectively capture particulates, gases, and odors, offering superior efficiency in high-moisture systems compared to standard air filters.
  • Specialized, 100% high-efficiency mechanical media captures particulates, gases, and odors
  • Heavy-duty, high wet-strength beverage board enclosure frame
  • Rust-resistant, expanded metal support grid for two-ply media
  • Carbon impregnated downstream layer absorbs VOCs and odors
  • Features 9 pleats per linear foot for high dust-holding capacity
  • Upstream layer with 70% efficiency for 3 to 10 micrometers
  • Maximum operating temperature of 150 degrees Fahrenheit
  • MERV-8 per ASHRAE 52.2-2017 standards
Catalog No. 50-005-06779 Supplier MSC Supplier No. 76935733
This extended surface carbon / particulate pleated panel filter is designed for superior filtration in commercial HVAC systems. It utilizes specialized, 100% high-efficiency mechanical media to effectively capture particulates, gases, and odors, offering superior efficiency in high-moisture systems compared to standard air filters.
  • Specialized, 100% high-efficiency mechanical media captures particulates, gases, and odors
  • Heavy-duty, high wet-strength beverage board enclosure frame
  • Rust-resistant, expanded metal support grid for two-ply media
  • Carbon impregnated downstream layer absorbs VOCs and odors
  • Features 9 pleats per linear foot for high dust-holding capacity
  • Upstream layer with 70% efficiency for 3 to 10 micrometers
  • Maximum operating temperature of 150 degrees Fahrenheit
  • MERV-8 per ASHRAE 52.2-2017 standards