Learn More
MSC Value Collection Fender Bumper, Synthetic Rubber, Half-Oval, 12 x 2 in
Catalog No. 5000532782
Encompass
This extruded half-oval fender bumper, made from synthetic rubber, is designed for various applications requiring reliable protection. Its D-shape profile and specific dimensions make it suitable for a range of uses.
  • Made from rubber material.
  • Features extruded construction for durability.
  • Designed with a D-shape for effective bumping.
  • Measures 12 inches wide and 2 inches high.
  • Includes a 0.39 inch bolt hole diameter for easy installation.
Catalog No. 50-005-32782 Supplier MSC Supplier No. 51380251
This extruded half-oval fender bumper, made from synthetic rubber, is designed for various applications requiring reliable protection. Its D-shape profile and specific dimensions make it suitable for a range of uses.
  • Made from rubber material.
  • Features extruded construction for durability.
  • Designed with a D-shape for effective bumping.
  • Measures 12 inches wide and 2 inches high.
  • Includes a 0.39 inch bolt hole diameter for easy installation.