Learn More
Waters Corp Waters Autosampler Vial, Polypropylene, 12 x 32 mm, Screw Neck, Preslit PTFE/Silicone Septum
Catalog No. 50818794 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This clean polypropylene vial offers a cost-effective alternative to glass, especially when compounds adhere to polar glass surfaces. Designed for analytical testing, it ensures efficiency and high throughput. The screw neck creates a tight seal, minimizing evaporation and contamination. A preslit PTFE/Silicone septum simplifies needle penetration, reduces coring, and ensures effective resealing. It is compatible with most autosamplers.
  • Clean polypropylene construction.
  • Cost-effective alternative to glass for polar-sensitive applications.
  • Tight screw neck seal minimizes evaporation and contamination.
  • Preslit PTFE/Silicone septum simplifies needle penetration, reduces coring, and ensures an airtight seal.
  • Compatible with most autosamplers.
  • Standardized 12 x 32 mm dimensions for seamless integration.
  • Offers exceptional chemical resistance.
Catalog No. 50-818-794 Supplier Waters Corp Supplier No. 186002639
This clean polypropylene vial offers a cost-effective alternative to glass, especially when compounds adhere to polar glass surfaces. Designed for analytical testing, it ensures efficiency and high throughput. The screw neck creates a tight seal, minimizing evaporation and contamination. A preslit PTFE/Silicone septum simplifies needle penetration, reduces coring, and ensures effective resealing. It is compatible with most autosamplers.
  • Clean polypropylene construction.
  • Cost-effective alternative to glass for polar-sensitive applications.
  • Tight screw neck seal minimizes evaporation and contamination.
  • Preslit PTFE/Silicone septum simplifies needle penetration, reduces coring, and ensures an airtight seal.
  • Compatible with most autosamplers.
  • Standardized 12 x 32 mm dimensions for seamless integration.
  • Offers exceptional chemical resistance.