Learn More
Waters Corp Waters Vial, LCGC Certified, Amber Glass, 12 x 32 mm
Catalog No. 50782540 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
Waters Certified vials are rigorously tested for cleanliness by HPLC and manufactured to precise dimensional tolerances. Designed for convenient storage of analytes and samples, they prevent deterioration and offer a secure fit with analytical equipment.
  • LCGC certified for cleanliness.
  • Amber glass construction protects against light-induced degradation.
  • Features a 12 x 32 mm screw neck design.
  • Includes a universal screw cap and PTFE/silicone septum.
  • Volume capacity of 300 μL.
  • Ideal for MS applications with excellent solvent resistance.
  • Compatible with autosamplers for efficient sample analysis.
Catalog No. 50-782-540 Supplier Waters Corp Supplier No. 186001130C
Waters Certified vials are rigorously tested for cleanliness by HPLC and manufactured to precise dimensional tolerances. Designed for convenient storage of analytes and samples, they prevent deterioration and offer a secure fit with analytical equipment.
  • LCGC certified for cleanliness.
  • Amber glass construction protects against light-induced degradation.
  • Features a 12 x 32 mm screw neck design.
  • Includes a universal screw cap and PTFE/silicone septum.
  • Volume capacity of 300 μL.
  • Ideal for MS applications with excellent solvent resistance.
  • Compatible with autosamplers for efficient sample analysis.