Learn More
Waters Corp Waters Vial, LCGC Certified, Amber Glass, 12 x 32 mm, Screw Neck
Catalog No. 50782542 Shop All Waters Chromatography Solutions Products
Encompass_Preferred
This LCGC Certified Amber Glass 12 x 32 mm Screw Neck Vial, Qsert, with Cap and Preslit PTFE/Silicone Septum, has a 300 μL volume. It is tested for cleanliness by HPLC and manufactured to tight dimensional tolerances, making it suitable for use with virtually all HPLC systems.
  • Vial comes with a cap and preslit PTFE/Silicone Septum for an excellent sealing mechanism.
  • Amber glass protects samples from UV light deterioration.
  • Chemical inertness of the LCGC certified amber glass ensures no interference with analysis.
  • Each batch is thoroughly UV-tested by HPLC to detect trace levels of chemicals.
  • PTFE/Silicone Septum is recommended for multiple injections and sample storage.
  • Septum offers excellent resealing characteristics and chemical resistance until punctured.
  • Recommended working temperature range for the PTFE/Silicone Septum is -40 °C to 200 °C.
Catalog No. 50-782-542 Supplier Waters Corp Supplier No. 186001131C
This LCGC Certified Amber Glass 12 x 32 mm Screw Neck Vial, Qsert, with Cap and Preslit PTFE/Silicone Septum, has a 300 μL volume. It is tested for cleanliness by HPLC and manufactured to tight dimensional tolerances, making it suitable for use with virtually all HPLC systems.
  • Vial comes with a cap and preslit PTFE/Silicone Septum for an excellent sealing mechanism.
  • Amber glass protects samples from UV light deterioration.
  • Chemical inertness of the LCGC certified amber glass ensures no interference with analysis.
  • Each batch is thoroughly UV-tested by HPLC to detect trace levels of chemicals.
  • PTFE/Silicone Septum is recommended for multiple injections and sample storage.
  • Septum offers excellent resealing characteristics and chemical resistance until punctured.
  • Recommended working temperature range for the PTFE/Silicone Septum is -40 °C to 200 °C.