Learn More
Waters Corp Waters Corporation Vial, LCGC Certified, Clear Glass, 12 x 32 mm, 1.5 mL, Screw Neck, Max Recovery
Catalog No. 50819158 Shop All Waters Corporation Products
Encompass_Preferred
These LCGC Certified clear glass vials are HPLC-tested for cleanliness and held to tight dimensional tolerances. Manufactured to ISO 9001 standards, they ensure autosampler compatibility and are certified for LC/MS analysis. Screw caps provide an excellent seal, and preslit PTFE/Silicone septa offer superior resealing for multiple injections and storage.
  • HPLC tested for cleanliness
  • Tight dimensional tolerances, ISO 9001 manufactured
  • Certified for critical dimensions: height, width, neck, threads, bottom thickness
  • UV-tested by HPLC for trace chemicals
  • Batch tested after packaging for cleanliness and quality
  • Screw caps provide excellent seal, holding septum during piercing
  • Preslit PTFE/Silicone septa offer excellent resealing
  • Recommended for multiple injections and sample storage
  • Working temperature range: -40°C to 200°C
Catalog No. 50-819-158 Supplier Waters Corp Supplier No. 186000327C
These LCGC Certified clear glass vials are HPLC-tested for cleanliness and held to tight dimensional tolerances. Manufactured to ISO 9001 standards, they ensure autosampler compatibility and are certified for LC/MS analysis. Screw caps provide an excellent seal, and preslit PTFE/Silicone septa offer superior resealing for multiple injections and storage.
  • HPLC tested for cleanliness
  • Tight dimensional tolerances, ISO 9001 manufactured
  • Certified for critical dimensions: height, width, neck, threads, bottom thickness
  • UV-tested by HPLC for trace chemicals
  • Batch tested after packaging for cleanliness and quality
  • Screw caps provide excellent seal, holding septum during piercing
  • Preslit PTFE/Silicone septa offer excellent resealing
  • Recommended for multiple injections and sample storage
  • Working temperature range: -40°C to 200°C