Learn More
Waters Corp Waters Vial, LCGC Certified, Amber Glass, 12 x 32 mm, 2 mL
Supplier: Waters Corp 186000847C
This LCGC Certified Amber Glass Screw Neck Vial, with Cap and Preslit PTFE/Silicone Septum, features a 2 mL volume and measures 12 x 32 mm. It is tested for cleanliness by HPLC and manufactured to the tightest dimensional tolerances in the industry.
- Vials are tested for cleanliness by HPLC and held to tightest dimensional tolerances in the industry.
- Certified.
- LCGC Certified.
- Autosampler vial.
- Includes vial, cap, and septum.
- Compatible with both liquid chromatography (LC) and gas chromatography (GC) techniques.
- Amber glass material is highly resistant to high temperatures, ensuring reliable performance during analysis.
- Certified for use in regulated environments.
- Amber color protects contents from UV light, which can damage certain chemicals.